missing translation for 'onlineSavingsMsg'
Få mere at vide

Carvedilol Biscarbazole (1,1'-[[2-(2-Methoxyphenoxy)ethyl]nitrilo]-bis[3-(9H-carbazol-4-yloxy)propan-2-ol]), LoGiCal

Artikelnummer. 30460792
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460792 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30460792 Leverandør LoGiCal Leverandørnr. LGCFOR0291.02

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Carvedilol Biscarbazole
CAS 918903-20-5
Analyt- eller komponentnavne Carvedilol Biscarbazole
Forsendelsestilstand Room Temperature
Molekylær formel C39 H39 N3 O6
Certificeringer/compliance ISO 9001
Synonym 1,1'-[[2-(2-Methoxyphenoxy)ethyl]nitrilo]bis[3-(9H-carbazol-4-yloxy)propan-2-ol], Carvedilol Imp. B (EP), Carvedilol Biscarbazole, Carvedilol USP Related Compound B, Carvedilol USP RC B, USP Carvedilol Related Compoud B, Ph Eur Carvedilol Impurity B, 1,1'-[2-(2-Methoxyphenoxy)ethyl]iminobis[3-(9H-carbazol-4-yloxy)-2-propanol]), SKF-105844, Carvedilol Related Compound B, Carvedilol Impurity B
InChI formel InChI=1S/C39H39N3O6/c1-45-34-16-6-7-17-35(34)46-21-20-42(22-26(43)24-47-36-18-8-14-32-38(36)28-10-2-4-12-30(28)40-32)23-27(44)25-48-37-19-9-15-33-39(37)29-11-3-5-13-31(29)41-33/h2-19,26-27,40-41,43-44H,20-25H2,1H3
Løsningstype Neat
SMIL COc1ccccc1OCCN(CC(O)COc2cccc3[nH]c4ccccc4c23)CC(O)COc5cccc6[nH]c7ccccc7c56
IUPAC navn 1-(9H-carbazol-4-yloxy)-3-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]-[2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol
Industry Type Clinical Testing
Molekylvægt (g/mol) 645.74
Mængde 10 mg
Formel vægt 645.2839 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.