Learn More
Applied Biosystems™ Carrier RNA
Ambion™ Carrier RNA (1 mg⁄mL) is a component of all the MagMAX™ Isolation Kits that may be purchased separately. Provided in one tube of 550 μL.
Brand:  Applied Biosystems™ 4382878
Additional Details : Weight : 0.10500kg
Product Code. 10127584
  • 1292.00 DKK / Each
In Stock 48
Add to basket
Specifications
Specifications
Store solution at –20°C.
Carrier RNA
Ambion™
Carrier for Nucleic Acid Precipitation
Dry Ice
Videos
Documents
For Research Use Only. Not for use in diagnostic procedures.