Få mere at vide
MedChemExpress Carperitide acetate
Artikelnummer. 16452859
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16452859 1 mg
1 options
Artikelnummer. 16452859 Leverandør MedChemExpress Leverandørnr. HYP1235A
MedChemExpress Carperitide acetate (Atrial Natriuretic Peptide (ANP) (1-28), human, porcine acetate) is a 28-amino acid hormone, that is normally produced and secreted by the human heart in response to cardiac injury and mechanical stretch. Carperitide acetate inhibits endothelin-1 secretion in a dose-dependent way.
Tekniske data
Renhed 96.81%
CAS 1366000-58-9
Indhold og opbevaring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Til brug med (applikation) COVID-19-immunoregulation
Molekylvægt (g/mol) 3140.5 g/mol
Produkttype Peptides
Mængde 1 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 50 mg/mL (15.92 mM∣Need ultrasonic)
Synonym Atrial Natriuretic Peptide (ANP) (1-28), human, porcine acetate