missing translation for 'onlineSavingsMsg'
Learn More

Carnosic Acid, TRC

Product Code. 17989004
Change view
Click to view available options
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17989004 5 mg
17908934 25 mg
17977954 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17989004 Supplier Toronto Research Chemicals Supplier No. TRCC1841805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3650-09-7
Molekylær formel C20 H28 O4
Molekylvægt (g/mol) 332.43
Synonym 4a(2H)-Phenanthrenecarboxylic acid, 1,3,4,9,10,10a-hexahydro-5,6-dihydroxy-1,1-dimethyl-7-(1-methylethyl)-, (4aR,10aS)-, 4a(2H)-Phenanthrenecarboxylic acid, 1,3,4,9,10,10a-hexahydro-5,6-dihydroxy-1,1-dimethyl-7-(1-methylethyl)-, (4aR-trans)-, Podocarpa-8,11,13-trien-17-oic acid, 11,12-dihydroxy-13-isopropyl- (7CI,8CI), (4aR,10aS)-1,3,4,9,10,10a-Hexahydro-5,6-dihydroxy-1,1-dimethyl-7-(1-methylethyl)-4a(2H)-phenanthrenecarboxylic acid, Carnosic acid, Rosamox, RoseOx, Salvin
IUPAC navn (4aR,10aS)-5,6-dihydroxy-1,1-dimethyl-7-propan-2-yl-2,3,4,9,10,10a-hexahydrophenanthrene-4a-carboxylic acid
SMIL CC(C)c1cc2CC[C@H]3C(C)(C)CCC[C@]3(C(=O)O)c2c(O)c1O

Specifications

Kemisk navn eller materiale Carnosic Acid
Mængde 5 mg
InChI formel InChI=1S/C20H28O4/c1-11(2)13-10-12-6-7-14-19(3,4)8-5-9-20(14,18(23)24)15(12)17(22)16(13)21/h10-11,14,21-22H,5-9H2,1-4H3,(H,23,24)/t14-,20+/m0/s1
Formel vægt 332.1988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.