Få mere at vide
Carlina Oxide, TRC
Artikelnummer. 30362356
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362356 100 mg
30353889 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362356 Leverandør Toronto Research Chemicals Leverandørnr. TRCC183600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 502-22-7
Molekylær formel C13 H10 O
Molekylvægt (g/mol) 182.218
Synonym 2-(3-Phenyl-1-propyn-1-yl)-furan, 2-(3-Phenyl-1-propynyl)-furan
IUPAC navn 2-(3-phenylprop-1-ynyl)furan
SMIL C(C#Cc1occc1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Carlina Oxide
Mængde 100 mg
InChI formel InChI=1S/C13H10O/c1-2-6-12(7-3-1)8-4-9-13-10-5-11-14-13/h1-3,5-7,10-11H,8H2
Formel vægt 182.073