missing translation for 'onlineSavingsMsg'
Learn More
Cariprazine, MedChemExpress Product Code.: 16446158

Cariprazine, MedChemExpress

Product Code. p-200099391
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446158

Brand: Medchem Express HY14763

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cariprazine is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).

Cariprazine is a novel antipsychotic drug candidate that exhibits high affinity for the D3 (Ki=0.085 nM) and D2 (Ki=0.49 nM) receptors, and moderate affinity for the 5-HT1A receptor (Ki=2.6 nM).

Specifications

Chemical Name or Material Cariprazine
Physical Form Solid
CAS 839712-12-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H32Cl2N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym RGH-188
Solubility Information DMSO : 2 mg/mL (4.68 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N[C@H]1CC[C@H](CCN2CCN(C3=CC=CC(Cl)=C3Cl)CC2)CC1)N(C)C
Molecular Weight (g/mol) 427.41
Formula Weight 427.41
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Cariprazine, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.