Få mere at vide
Cariprazine-d8, TRC
Artikelnummer. 30358149
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30358149 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358149 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1834922.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cariprazine-d8
CAS 1308278-69-4
Renhedsgrad noter HPLC
Molekylær formel C21 D8 H24 Cl2 N4 O
Mængde 2.5 mg
Synonym Urea, N'-[trans-4-[2-[4-(2,3-dichlorophenyl)-1-piperazinyl]ethyl-2,2-d2]cyclohexyl]-N,N-di(methyl-d3), N'-[trans-4-[2-[4-(2,3-Dichlorophenyl)-1-piperazinyl]ethyl-2,2-d2]cyclohexyl]-N,N-di(methyl-d3)-urea
InChI formel InChI=1S/C21H32Cl2N4O/c1-25(2)21(28)24-17-8-6-16(7-9-17)10-11-26-12-14-27(15-13-26)19-5-3-4-18(22)20(19)23/h3-5,16-17H,6-15H2,1-2H3,(H,24,28)/t16-,17-/i1D3,2D3,11D2
SMIL [2H]C([2H])([2H])N(C(=O)N[C@@H]1CC[C@@H](CC([2H])([2H])N2CCN(CC2)c3cccc(Cl)c3Cl)CC1)C([2H])([2H])[2H]
IUPAC navn 3-[4-[2,2-dideuterio-2-[4-(2,3-dichlorophenyl)piperazin-1-yl]ethyl]cyclohexyl]-1,1-bis(trideuteriomethyl)urea
Molekylvægt (g/mol) 435.46
Formel vægt 434.2455
Procent renhed >95