Få mere at vide
Carfentrazone Ethyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carfentrazone-ethyl |
| CAS | 128639-02-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C15 H14 Cl2 F3 N3 O3 |
| Mængde | 100 mg |
| Synonym | Aim, Aim (herbicide), Aurora, Aurora (pesticide), Aurora 50WG, Carfentrazone-ethyl, Ethyl (RS)-2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorophenyl]propionate, F 8426, Kuaimieling, Quicksilver T&O, Shark EW, Spotlight, Spotlight (triazolinone herbicide), Spotlight 24EC, Spotlight Plus, Task, Task (herbicide), Zuocaotong |
| InChI formel | InChI=1S/C15H14Cl2F3N3O3/c1-3-26-13(24)10(17)4-8-5-12(11(18)6-9(8)16)23-15(25)22(14(19)20)7(2)21-23/h5-6,10,14H,3-4H2,1-2H3 |
| SMIL | CCOC(=O)C(Cl)Cc1cc(N2N=C(C)N(C(F)F)C2=O)c(F)cc1Cl |
| IUPAC navn | ethyl 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoate |
| Molekylvægt (g/mol) | 412.19 |
| Vis mere |