missing translation for 'onlineSavingsMsg'
Få mere at vide

Carfentrazone Ethyl Ester, TRC

Artikelnummer. 30356559
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356559 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30356559 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18345525MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Carfentrazone-ethyl
CAS 128639-02-1
Renhedsgrad noter HPLC
Molekylær formel C15 H14 Cl2 F3 N3 O3
Mængde 25 mg
Synonym Aim, Aim (herbicide), Aurora, Aurora (pesticide), Aurora 50WG, Carfentrazone-ethyl, Ethyl (RS)-2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-4,5-dihydro-3-methyl-5-oxo-1H-1,2,4-triazol-1-yl]-4-fluorophenyl]propionate, F 8426, Kuaimieling, Quicksilver T&O, Shark EW, Spotlight, Spotlight (triazolinone herbicide), Spotlight 24EC, Spotlight Plus, Task, Task (herbicide), Zuocaotong
InChI formel InChI=1S/C15H14Cl2F3N3O3/c1-3-26-13(24)10(17)4-8-5-12(11(18)6-9(8)16)23-15(25)22(14(19)20)7(2)21-23/h5-6,10,14H,3-4H2,1-2H3
SMIL CCOC(=O)C(Cl)Cc1cc(N2N=C(C)N(C(F)F)C2=O)c(F)cc1Cl
IUPAC navn ethyl 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoate
Molekylvægt (g/mol) 412.19
Formel vægt 411.0364
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.