Få mere at vide
Carfentrazone Ethyl-d5, TRC
Artikelnummer. 30361721
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30361721 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361721 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18345625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carfentrazone Ethyl D5 (ethyl D5)
CAS 2140327-60-0
Molekylær formel C15 D5 H9 Cl2 F3 N3 O3
Mængde 25 mg
Synonym Carfentrazone Ethyl D5 (ethyl D5), Carfentrazone Ethyl-d5, Carfentrazone Ethyl D5 (ethyl D5)
InChI formel InChI=1S/C15H14Cl2F3N3O3/c1-3-26-13(24)10(17)4-8-5-12(11(18)6-9(8)16)23-15(25)22(14(19)20)7(2)21-23/h5-6,10,14H,3-4H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)C(Cl)Cc1cc(N2N=C(C)N(C(F)F)C2=O)c(F)cc1Cl
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoate
Molekylvægt (g/mol) 417.222
Formel vægt 416.068