missing translation for 'onlineSavingsMsg'
Få mere at vide

Carfentrazone Ethyl-d5, TRC

Artikelnummer. 30360671
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360671 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360671 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18345650MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Carfentrazone Ethyl D5 (ethyl D5)
CAS 2140327-60-0
Molekylær formel C15 D5 H9 Cl2 F3 N3 O3
Mængde 50 mg
Synonym Carfentrazone Ethyl D5 (ethyl D5), Carfentrazone Ethyl-d5, Carfentrazone Ethyl D5 (ethyl D5)
InChI formel InChI=1S/C15H14Cl2F3N3O3/c1-3-26-13(24)10(17)4-8-5-12(11(18)6-9(8)16)23-15(25)22(14(19)20)7(2)21-23/h5-6,10,14H,3-4H2,1-2H3/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)C(Cl)Cc1cc(N2N=C(C)N(C(F)F)C2=O)c(F)cc1Cl
IUPAC navn 1,1,2,2,2-pentadeuterioethyl 2-chloro-3-[2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-1,2,4-triazol-1-yl]-4-fluorophenyl]propanoate
Molekylvægt (g/mol) 417.222
Formel vægt 416.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.