Få mere at vide
Tocris Bioscience™ Cardamonin
Artikelnummer. 18074042
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18074042 50 mg
18034812 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18074042 Leverandør Tocris Bioscience™ Leverandørnr. 2509/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Inhibitor of NF-κB activation; anti-inflammatory
Chalone analog that display anti-inflammatory activity. Inhibits NO and PGE2 production from LPS- and IFN-γ-induced RAW cells and inhibits TXB2 production via the COX-1 and COX-2 pathways. Inhibits NF-κB activation via inhibition of IκBα degradation and phosphorylation, IκB kinase activation and NF-κB nuclear translocation.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19309-14-9
Molekylær formel C16H14O4
Tekniske data
Kemisk navn eller materiale Cardamonin
Mængde 50 mg
Formel vægt 270.28
Renhed >98%
Molekylvægt (g/mol) 270.284