missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Carbuterol-D9 Hemisulfate Salt, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carbuterol-d9 Hemisulfate Salt |
| CAS | 1346747-24-7 |
| Molekylær formel | C13H12D9N3O3 • 1/2 H2SO4 |
| Mængde | 10 mg |
| Synonym | N-[5-[2-[(1,1-Dimethylethyl-9)amino]-1-hydroxyethyl]-2-hydroxyphenyl]urea, Hemisulfate Salt, Bronsecur-d9 Hemisulfate Salt, Carbuterol-d9 Hemisulfate Salt, Pirem-d9 Hemisulfate Salt, SKF 40383-d9 Hemisulfate Salt, SKF 40383A-d9 Hemisulfate Salt |
| InChI formel | InChI=1S/C13H21N3O3/c1-13(2,3)15-7-11(18)8-4-5-10(17)9(6-8)16-12(14)19/h4-6,11,15,17-18H,7H2,1-3H3,(H3,14,16,19)/i1D3,2D3,3D3 |
| SMIL | OC1=CC=C(C(O)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1NC(N)=O |
| IUPAC navn | [5-[2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-1-hydroxyethyl]-2-hydroxyphenyl]urea |
| Molekylvægt (g/mol) | 276.384904 |
| Formel vægt | 276.2148 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?