Få mere at vide
Carbutamide-d9, TRC
Artikelnummer. 30350609
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30350609 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350609 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1833021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbutamide-d9
CAS 1246820-50-7
Molekylær formel C11H8D9N3O3S
Mængde 1 mg
Synonym 4-Amino-N-[(butylamino-d9)carbonyl]benzenesulfonamide, N1-(Butyl-d9-carbamoyl)sulfanilamide, 1-(Butyl-d9)-3-sulfanilylurea, Alentin-d9, Aminophenurobutane-d9, BZ 55-d9, Bucarban-d9, Bucrol-d9, Bukarban-d9, Burcol-d9, Butisulfina-d9, Carbutamid-d9, Diaboral-d9, Emedan-d9, Glucidoral-d9, Glucofren-d9, Glybutamide-d9, Inbuton-d9, Invenol-d9, Oranil-d9, Oranyl-d9, Orasulin-d9, U 6987-d9
InChI formel InChI=1S/C11H17N3O3S/c1-2-3-8-13-11(15)14-18(16,17)10-6-4-9(12)5-7-10/h4-7H,2-3,8,12H2,1H3,(H2,13,14,15)/i1D3,2D2,3D2,8D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC(=O)NS(=O)(=O)c1ccc(N)cc1
IUPAC navn 1-(4-aminophenyl)sulfonyl-3-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)urea
Molekylvægt (g/mol) 280.39
Formel vægt 280.1556