missing translation for 'onlineSavingsMsg'
Få mere at vide

Carboxyphosphamide Benzyl Ester-d4, TRC

Artikelnummer. 30362981
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362981 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362981 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1813575MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Carboxyphosphamide Benzyl Ester-d4
CAS 1276302-73-8
Molekylær formel C14H17D4Cl2N2O4P
Mængde 5 mg
Synonym 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Phenylmethyl Ester-d4, 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Benzyl Ester-d4
SMIL [2H]C([2H])(Cl)CN(CC([2H])([2H])Cl)P(=O)(N)OCCC(=O)OCc1ccccc1
Molekylvægt (g/mol) 387.23
Formel vægt 386.0867
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.