missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Carboxypeptidase M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586657
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Purity is > 95% by SDS-PAGE.
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Tekniske data
| Carboxypeptidase M | |
| Polyclonal | |
| Unconjugated | |
| CPM | |
| 1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1368, 314855, 70574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14384, Q80V42 | |
| CPM | |
| Synthetic peptide, corresponding to amino acids 80-130 of Human CPM. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion