missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Carboxypeptidase M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551730
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: LKTVAQNYSS VTHLHSIGKS VKGRNLWVLV VGRFPKEHRI GIPEFKYVAN MHGDETVGRE LLLHLIDYLV TSDGKDPEIT NLINSTRIHI MPSMNPDGFE AVKKPDCYYS IGRENYNQYD LNRNFPDAFE YNNVS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Tekniske data
| Carboxypeptidase M | |
| Polyclonal | |
| Unconjugated | |
| CPM | |
| 1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1368 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14384 | |
| CPM | |
| Recombinant protein corresponding to Human Carboxypeptidase M. Recombinant protein control fragment (Product #RP-101018). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion