missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Carboxypeptidase M Antibody (105), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89359G
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Carboxypeptidase M Monoclonal specifically detects Carboxypeptidase M in Human samples. It is validated for Western Blot, ELISA.
Tekniske data
| Carboxypeptidase M | |
| Monoclonal | |
| DyLight 488 | |
| carboxypeptidase M, EC 3.4.17, EC 3.4.17.12 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxypeptidase M/ CPM (NP_938079.1; Met 1-His 422). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 105 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1368 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion