missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B2/CPB2 Antibody (173), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP2-90754V
Additional Details : Weight : 0.00970kg
Description
Carboxypeptidase B2/CPB2 Monoclonal specifically detects Carboxypeptidase B2/CPB2 in Mouse samples. It is validated for ELISA.Specifications
Carboxypeptidase B2/CPB2 | |
Monoclonal | |
DyLight 405 | |
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase B2/CPB2 (Q9JHH6; Met1-Thr422). | |
0.1 mL | |
Lipid and Metabolism | |
1361 | |
Mouse | |
Purified |
ELISA | |
173 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CPA1 Human CPB1 Mouse CPA1 Mouse CPB1 | |
Store at 4°C in the dark. | |
IgG |