Få mere at vide
Carboxymethyl Chitosan, TRC
Artikelnummer. 30349013
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349013 5 g
30357407 1 g
30365457 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30349013 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1811535G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83512-85-0
Molekylær formel (C6H26N2O10)n
Molekylvægt (g/mol) 543.5
Synonym Chitosan N-(carboxymethyl), Carboxymethylchitosan, N-Carboxymethylchitosan
IUPAC navn 2-[[(2R,3R,5S,6R)-5-[(2R,3R,5S,6R)-3-(carboxymethylamino)-6-(hydroxymethyl)-5-methoxyoxan-2-yl]oxy-2-hydroxy-6-(hydroxymethyl)oxan-3-yl]amino]acetic acid
SMIL CO[C@H]1C[C@@H](NCC(=O)O)[C@H](O[C@H]2C[C@@H](NCC(=O)O)[C@H](O)O[C@@H]2CO)O[C@@H]1CO
Tekniske data
Kemisk navn eller materiale Carboxymethyl Chitosan
Mængde 5 g
InChI formel InChI=1S/C17H30N2O11/c1-27-10-3-9(19-5-15(24)25)17(30-12(10)6-20)29-11-2-8(18-4-14(22)23)16(26)28-13(11)7-21/h8-13,16-21,26H,2-7H2,1H3,(H,22,23)(H,24,25)/t8-,9-,10+,11+,12-,13-,16-,17-/m1/s1
Formel vægt 438.185