Få mere at vide
Carboxymefloquine, TRC
Artikelnummer. 30368300
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368300 50 mg
30360882 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368300 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17870050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35853-50-0
Molekylær formel C12 H5 F6 N O2
Molekylvægt (g/mol) 309.16
Synonym 4-Quinolinecarboxylic acid, 2,8-bis(trifluoromethyl)-Ro 21-5104
IUPAC navn 2,8-bis(trifluoromethyl)quinoline-4-carboxylic acid
SMIL OC(=O)c1cc(nc2c(cccc12)C(F)(F)F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Carboxymefloquine
Mængde 50 mg
InChI formel InChI=1S/C12H5F6NO2/c13-11(14,15)7-3-1-2-5-6(10(20)21)4-8(12(16,17)18)19-9(5)7/h1-4H,(H,20,21)
Formel vægt 309.0224