missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxylesterase 3/CES3/Esterase 31 Antibody (033), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP2-89802H
Additional Details : Weight : 0.00970kg
Description
Carboxylesterase 3/CES3/Esterase 31 Monoclonal specifically detects Carboxylesterase 3/CES3/Esterase 31 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Carboxylesterase 3/CES3/Esterase 31 | |
Monoclonal | |
HRP | |
carboxylesterase 3, carboxylesterase 3 (brain), EC 3.1.1, EC 3.1.1.1, ES31FLJ21736, esterase 31, Liver carboxylesterase 31 homolog | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carboxylesterase 3/CES3/Esterase 31 (NP_079198.2; Met1-Glu561). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
33 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
23491 | |
Store at 4°C in the dark. | |
IgG |