missing translation for 'onlineSavingsMsg'
Få mere at vide

Carboxin Sulfoxide, TRC

Artikelnummer. 30137781
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137781 250 mg
30136411 500 mg
17934495 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30137781 Leverandør Toronto Research Chemicals Leverandørnr. TRCC177605250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17757-70-9
Molekylær formel C12 H13 N O3 S
Molekylvægt (g/mol) 251.3
Synonym Carboxin-sulfoxide, 1,4-Oxathiin-3-carboxanilide, 5,6-dihydro-2-methyl-, 4-oxide (8CI), 5,6-Dihydro-2-methyl-1,4-oxathiin-3-carboxanilide-4-oxide, 5,6-Dihydro-3-carboxanilide-2-methyl-1,4-oxathiin-4-oxide, Carboxin 4-oxide, Carboxin sulfoxide, F 831, Vitavax sulfoxide
IUPAC navn 6-methyl-4-oxo-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamide
SMIL CC1=C(C(=O)Nc2ccccc2)S(=O)CCO1

Tekniske data

Kemisk navn eller materiale Carboxin-sulfoxide
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C12H13NO3S/c1-9-11(17(15)8-7-16-9)12(14)13-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,13,14)
Formel vægt 251.0616
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.