Få mere at vide
Carbophenothion, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carbophenothion |
| CAS | 786-19-6 |
| Molekylær formel | C11 H16 Cl O2 P S3 |
| Mængde | 250 mg |
| Synonym | Phosphorodithioic acid, S-[[(p-chlorophenyl)thio]methyl] O,O-diethyl ester (6CI,8CI), Acarithion, Carbofenothion, Carbofenotion, Carbofenthion, Carbophenothion, ENT 23,708, Ethyl carbophenothion, Garrathion, Hexathion, NSC 231691, O,O-Diethyl S-(p-chlorophenylthio)methyl phosphorodithioate, Oleoakarithion, R 1303, R 1303 (pesticide), S-[[(p-Chlorophenyl)thio]methyl] O,O-diethyl phosphorodithioate, Stauffer R 1303, Trithion |
| InChI formel | InChI=1S/C11H16ClO2PS3/c1-3-13-15(16,14-4-2)18-9-17-11-7-5-10(12)6-8-11/h5-8H,3-4,9H2,1-2H3 |
| SMIL | CCOP(=S)(OCC)SCSc1ccc(Cl)cc1 |
| IUPAC navn | (4-chlorophenyl)sulfanylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 342.87 |
| Formel vægt | 341.9739 |