missing translation for 'onlineSavingsMsg'
Få mere at vide

Carbophenothion-methyl 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Artikelnummer. 30328276
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328276 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30328276 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA11020500AL100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale Carbophenothion-methyl
CAS 953-17-3
Analyt- eller komponentnavne Carbophenothion-methyl
Forsendelsestilstand Room Temperature
Molekylær formel C9 H12 Cl O2 P S3
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym Phosphorodithioic acid, S-[[(4-chlorophenyl)thio]methyl] O,O-dimethyl ester, Phosphorodithioic acid, S-[[(p-chlorophenyl)thio]methyl] O,O-dimethyl ester (6CI,8CI), Carbophenothion methyl, Methyl carbophenothion, Methyl trithion, NSC 231692, S-[[(p-Chlorophenyl)thio]methyl] O,O-dimethyl phosphorodithioate, Stauffer R 1492, Trithion-methyl, Carbophenothion-methyl
InChI formel InChI=1S/C9H12ClO2PS3/c1-11-13(14,12-2)16-7-15-9-5-3-8(10)4-6-9/h3-6H,7H2,1-2H3
SMIL COP(=S)(OC)SCSc1ccc(Cl)cc1
IUPAC navn (4-chlorophenyl)sulfanylmethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 314.81
Mængde 1 ml
Formel vægt 313.9426 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.