Få mere at vide
Carbonochloridic Acid Octadecyl Ester, TRC
Artikelnummer. 30363768
Skift visning
Mængde:
2.5 g
Artikelnummer. Mængde
30363768 2.5 g
1 options
Artikelnummer. 30363768 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1768702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbonochloridic Acid Octadecyl Ester
CAS 51637-93-5
Molekylær formel C19H37ClO2
Mængde 2.5 g
Synonym Chloro-formic Acid Octadecyl Ester, Octadecyl Chloroformate, Stearyl Chloroformate, Stearyloxycarbonyl Chloride, n-Octadecyl Chloroformate
InChI formel InChI=1S/C19H37ClO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-19(20)21/h2-18H2,1H3
SMIL CCCCCCCCCCCCCCCCCCOC(=O)Cl
IUPAC navn octadecyl carbonochloridate
Molekylvægt (g/mol) 332.95