Få mere at vide
Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA537845
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Carbonic Anhydrase XII is a protein coding gene.
Tekniske data
| Carbonic Anhydrase XII | |
| Polyclonal | |
| Unconjugated | |
| CA12 | |
| 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43570 | |
| CA12 | |
| Peptide with sequence C-SHLQHVKYKGQE, from the internal region of CA12 (aa188-199). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |