missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Carbonic Anhydrase VI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101493
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Antibody detects endogenous levels of total CA6.
The protein encoded by this gene is one of several isozymes of carbonic anhydrase. This protein is found only in salivary glands and saliva and protein may play a role in the reversible hydratation of carbon dioxide though its function in saliva is unknown.
Tekniske data
| Carbonic Anhydrase VI | |
| Polyclonal | |
| Unconjugated | |
| CA6 | |
| CA6; CA-6; Car6; Carbonate dehydratase VI; carbonic anhydrase 6; Carbonic anhydrase VI; carbonic anhydrase VI nirs variant 2; Carbonic anhydrase6; CA-VI; DOC1; GUSTIN; salivary carbonic anhydrase; Secreted carbonic anhydrase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 765 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23280 | |
| CA6 | |
| A synthesized peptide derived from human CA6(Accession P23280), corresponding to amino acid residues V237-S287. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion