Learn More
Invitrogen™ Carbonic Anhydrase VB Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5120567
Product Code. 17771964
  • 3735.00 DKK / 100µL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This item is not returnable. View return policy
Description
Description
Positive test controls include: A-549, Mouse kidney, Mouse lung, Mouse spleen, Rat lung. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MVVMNSLRVI LQASPGKLLW RKFQIPRFMP ARPCSLYTCT YKTRNRALHP LWESVDLVPG GDRQSPINIR WRDSVYDPGL KPLTISYDPA TCLHVWNNGY SFLVEFEDST DKSVIKGGPL EHNYRLKQFH FHWGAIDAWG SEHTVDSKCF PAELHLVHWN AVRFENFEDA ALEENGLAVI GVFLKLGKHH KELQKLVDTL PSIKHKDALV EFGSFDPSCL MPTCPDYWTY SGSLTTPPLS ESVTWIIKKQ PVEVDHDQLE QFRTLLFTSE GEKEKRMVDN FRPLQPLMNR TVRSSFRHDY VLNVQAKPKP ATSQATP.
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predominantly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase VB
Polyclonal
Unconjugated
CA5B
7330410H16Rik; Ca5b; Car5b; Carbonate dehydratase VB; carbonic anhydrase 5B; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase; CarVb; CAVB; CA-VB; D730005F19Rik
Rabbit
Affinity Chromatography
RUO
11238, 302669, 56078
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
0.18 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q66HG6, Q9QZA0, Q9Y2D0
CA5B, Car5b
Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human CA5B (NP_009151.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions