missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Carbonic Anhydrase IV Polyclonal Antibody

Beskrivelse
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CA4 is observed and less than 5% cross-reactivity with recombinant human CA1, 2, 3, 8, 9, 10, 12, and 13 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.
Tekniske data
Tekniske data
| Antigen | Carbonic Anhydrase IV |
| Applikationer | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 0.2 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 5% trehalose and No Preservative |
| Gene | CA4 |
| Genadgangsnr. | P22748 |
| Gene Alias | AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 |
| Gensymboler | CA4 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?