missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody

Beskrivelse
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Carbonic Anhydrase I, III, and XIII is observed and less than 1% cross-reactivity with rhCarbonic Anhydrase IV, VIII, IX, X, XII, and XIV is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Tekniske data
Tekniske data
| Antigen | Carbonic Anhydrase II |
| Applikationer | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 0.2 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 5% trehalose and No Preservative |
| Gene | CA2 |
| Genadgangsnr. | P00918 |
| Gene Alias | AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 |
| Gensymboler | CA2 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?