missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Carbonic Anhydrase II Polyclonal Antibody

Artikelnummer. 13462377
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
13462377 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 13462377 Leverandør Invitrogen™ Leverandørnr. PA528267

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Dog (92%), Rabbit (89%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Carbonic Anhydrase II
Applikationer Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Koncentration 0.17 mg/mL
Konjugeret Unconjugated
Formulering PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CA2
Genadgangsnr. P00918, P00920
Gene Alias AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5
Gensymboler CA2, Car2
Værtsarter Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Carbonic anhydrase II. The exact sequence is proprietary.
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 12349, 760
Målarter Human, Mouse
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.