missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Carbonic Anhydrase II/CA2 Antibody (002), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90676IR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Carbonic Anhydrase II/CA2 Monoclonal specifically detects Carbonic Anhydrase II/CA2 in Mouse samples. It is validated for ELISA.
Tekniske data
| Carbonic Anhydrase II/CA2 | |
| Monoclonal | |
| DyLight 755 | |
| CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase II/CA2 (AAH55291.1; Met 1-Lys 260). | |
| 0.1 mL | |
| Hypoxia, Lipid and Metabolism | |
| 760 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 2 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion