missing translation for 'onlineSavingsMsg'
Få mere at vide

Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester, TRC

Artikelnummer. 30362723
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362723 500 mg
30361727 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30362723 Leverandør Toronto Research Chemicals Leverandørnr. TRCC176635500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918822-70-5
Molekylær formel C9H11NO7S
Molekylvægt (g/mol) 277.25
Synonym Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester, [[2-(Ethenylsulfonyl)ethoxy]carbonyl]oxy]-2,5-pyrrolidinedione, Succinimidyl 2-(Vinylsulfonyl)ethyl Carbonate, SVEC
IUPAC navn (2,5-dioxopyrrolidin-1-yl) 2-ethenylsulfonylethyl carbonate
SMIL C=CS(=O)(=O)CCOC(=O)ON1C(=O)CCC1=O

Specifications

Kemisk navn eller materiale Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester
Mængde 500 mg
InChI formel InChI=1S/C9H11NO7S/c1-2-18(14,15)6-5-16-9(13)17-10-7(11)3-4-8(10)12/h2H,1,3-6H2
Formel vægt 277.0256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.