Få mere at vide
Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester, TRC
Artikelnummer. 30361727
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361727 50 mg
30362723 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361727 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17663550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 918822-70-5
Molekylær formel C9H11NO7S
Molekylvægt (g/mol) 277.25
Synonym Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester, [[2-(Ethenylsulfonyl)ethoxy]carbonyl]oxy]-2,5-pyrrolidinedione, Succinimidyl 2-(Vinylsulfonyl)ethyl Carbonate, SVEC
IUPAC navn (2,5-dioxopyrrolidin-1-yl) 2-ethenylsulfonylethyl carbonate
SMIL C=CS(=O)(=O)CCOC(=O)ON1C(=O)CCC1=O
Tekniske data
Kemisk navn eller materiale Carbonic Acid 2,5-Dioxo-1-pyrrolidinyl 2-(Ethenylsulfonyl)ethyl Ester
Mængde 50 mg
InChI formel InChI=1S/C9H11NO7S/c1-2-18(14,15)6-5-16-9(13)17-10-7(11)3-4-8(10)12/h2H,1,3-6H2
Formel vægt 277.0256