Få mere at vide
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 23
Mængde:
100/kasse
Beskrivelse
Leaf-shaped blade sharpened along its leading edge.
- Leaf-shaped.
- Sharpened along its leading edge.
- Used for making long incisions such as an upper midline incision of the abdomen during the repair of a perforated gastric ulcer.
- Fits handles 4, 4L, 4 graduated, and 6B.
Tekniske data
Tekniske data
| Bladmateriale | Kulstofstål |
| Sterilitet | Ikke steril |
| Mængde | 100/kasse |
| Emballagetype | Individuelt pakket |