Få mere at vide
Swann-Morton™ Carbon Steel Individual Foil Packs, Sterile Scalpel Blades, Major X
Artikelnummer. 15951352
Skift visning
Mængde:
50/æske
Artikelnummer. Mængde
15951352 50/æske
1 options
Artikelnummer. 15951352 Leverandør Swann-Morton™ Leverandørnr. 0726
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Thicker carbon steel for added strength.
  • Thicker specification of carbon steel.
  • Provides orthopedic surgeon with added strength when working close to bone and within joints.
  • Fits both the standard and long major handles.
Tekniske data
Sterilitet Sterile
Mængde 50/æske
Emballagetype Individually Packaged