Få mere at vide
Carbomethoxyhydrazide-d3, TRC
Artikelnummer. 30366530
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30366530 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366530 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17661725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbomethoxyhydrazide-d3
CAS 1794752-06-9
Molekylær formel C2H3D3N2O2
Mængde 25 mg
Synonym (Methoxycarbonyl)hydrazide-d3, (Methoxycarbonyl)hydrazine-d3, Carbomethoxyhydrazide-d3, Carbomethoxyhydrazine-d3, Methyl Carbazate-d3, Methyl Carbazinate-d3, Methyl Hydrazinecarboxylate-d3, Methyl Hydrazinocarboxylate-d3, Methyl Hydrazinoformate-d3, NSC 11709-d3
InChI formel InChI=1S/C2H6N2O2/c1-6-2(5)4-3/h3H2,1H3,(H,4,5)/i1D3
SMIL [2H]C([2H])([2H])OC(=O)NN
IUPAC navn trideuteriomethyl N-aminocarbamate
Molekylvægt (g/mol) 93.1
Formel vægt 93.0618