Få mere at vide
Carbofuran, TRC
Artikelnummer. 30360164
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360164 100 mg
30361267 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360164 Leverandør Toronto Research Chemicals Leverandørnr. TRCC176590100MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1563-66-2
Molekylær formel C12 H15 N O3
Molekylvægt (g/mol) 221.25
Synonym 7-Benzofuranol, 2,3-dihydro-2,2-dimethyl-, methylcarbamate (9CI), Carbamic acid, methyl-, 2,3-dihydro-2,2-dimethyl-7-benzofuranyl ester (8CI), 2,2-Dimethyl-2,3-dihydro-7-benzofuranyl N-methylcarbamate, 2,2-Dimethyl-7-coumaranyl N-methylcarbamate, 2,3-Dihydro-2,2-dimethyl-7-benzofuranol methylcarbamate, 2,3-Dihydro-2,2-dimethyl-7-benzofuranyl N-methylcarbamate, 2,3-Dihydro-2,2-dimethyl-7-benzofuranyl methylcarbamate, Adifur, BAY 70143, Brifer, Carbofuran, Chinufur, Curaterr, Curaterr 5, Diafuran, Diafuran 5G, Furadan, Furadan 350S, Furadan 350SC, Furadan 3G, Furadan 4F, Furadan 75WP, Furadan F, Furadan G, Furadan SP 50, NIA 10242, NSC 167822, Niagara 10242, OMS 864, Sunfuran, Terafuran, Yaltox
IUPAC navn (2,2-dimethyl-3H-1-benzofuran-7-yl) N-methylcarbamate
SMIL CNC(=O)Oc1cccc2CC(C)(C)Oc12
Tekniske data
Kemisk navn eller materiale Carbofuran
Mængde 100 mg
InChI formel InChI=1S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6H,7H2,1-3H3,(H,13,14)
Formel vægt 221.1052