Få mere at vide
Carbodenafil, LoGiCal
Artikelnummer. 30459633
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459633 10 mg
1 options
Artikelnummer. 30459633 Leverandør LoGiCal Leverandørnr. LGCFOR0254.38
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbodenafil
CAS 944241-52-5
Analyt- eller komponentnavne Carbodenafil
Forsendelsestilstand Room Temperature
Molekylær formel C24 H32 N6 O3
Certificeringer/compliance ISO 9001
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-ethyl-1-piperazinyl)carbonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Carbodenafil, Fondenafil
InChI formel InChI=1S/C24H32N6O3/c1-5-8-18-20-21(28(4)27-18)23(31)26-22(25-20)17-15-16(9-10-19(17)33-7-3)24(32)30-13-11-29(6-2)12-14-30/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,31)
Løsningstype Neat
SMIL CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)N4CCN(CC)CC4
IUPAC navn 5-[2-ethoxy-5-(4-ethylpiperazine-1-carbonyl)phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Industry Type Food Testing
Molekylvægt (g/mol) 452.55
Mængde 10 mg
Formel vægt 452.2536 g/mol