Få mere at vide
Carbocisteine Diethyl Ester Hydrochloride, Mikromol™
Artikelnummer. 17919774
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17919774 25 mg
1 options
Artikelnummer. 17919774 Leverandør Mikromol Leverandørnr. MM0236.130025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Carbocisteine Diethyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbocisteine Diethyl Ester Hydrochloride
CAS 92282-33-2
Analyt- eller komponentnavne Carbocisteine Diethyl Ester Hydrochloride
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C9 H17 N O4 S . Cl H
Certificeringer/compliance ISO 9001
Synonym Alanine, 3-[(carboxymethyl)thio]-, diethyl ester, hydrochloride (7CI), Carbocisteine Diethyl Ester Hydrochloride
InChI formel InChI=1S/C9H17NO4S.ClH/c1-3-13-8(11)6-15-5-7(10)9(12)14-4-2;/h7H,3-6,10H2,1-2H3;1H/t7-;/m0./s1
SMIL Cl.CCOC(=O)CSC[C@H](N)C(=O)OCC
IUPAC navn ethyl (2R)-2-amino-3-(2-ethoxy-2-oxoethyl)sulfanylpropanoate;hydrochloride
Molekylvægt (g/mol) 271.76
Mængde 25 mg
Formel vægt 271.0645 g/mol