Få mere at vide
Carbiphene Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carbiphene Hydrochloride |
| CAS | 467-22-1 |
| Analyt- eller komponentnavne | Carbiphene Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C28 H34 N2 O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Etomide Hydrochloride, Carbiphene Hydrochloride, Acetamide, 2-ethoxy-N-methyl-N-[2-(methylphenethylamino)ethyl]-2,2-diphenyl-, hydrochloride, Acetamide, 2-ethoxy-N-methyl-N-[2-(methylphenethylamino)ethyl]-2,2-diphenyl-, monohydrochloride, Benzeneacetamide, α-ethoxy-N-methyl-N-[2-[methyl(2-phenylethyl)amino]ethyl]-α-phenyl-, monohydrochloride, 2-Ethoxy-N-methyl-N-[2-(methylphenethylamino)ethyl]-2,2-diphenylacetamide hydrochloride, Bandol, Carbiphene hydrochloride, Etomide hydrochloride, Jubalon, NSC 106959, SQ 10269, Benzeneacetamide, α-ethoxy-N-methyl-N-[2-[methyl(2-phenylethyl)amino]ethyl]-α-phenyl-, hydrochloride (1:1) |
| InChI formel | InChI=1S/C28H34N2O2.ClH/c1-4-32-28(25-16-10-6-11-17-25,26-18-12-7-13-19-26)27(31)30(3)23-22-29(2)21-20-24-14-8-5-9-15-24;/h5-19H,4,20-23H2,1-3H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCOC(C(=O)N(C)CCN(C)CCc1ccccc1)(c2ccccc2)c3ccccc3 |
| Vis mere |