Få mere at vide
Carbazomycin A, TRC
Artikelnummer. 30357733
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357733 50 mg
30351723 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357733 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17583050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75139-39-8
Molekylær formel C16H17NO2
Molekylvægt (g/mol) 255.31
Synonym 3,4-Dimethoxy-1,2-dimethyl-9H-carbazole
IUPAC navn 3,4-dimethoxy-1,2-dimethyl-9H-carbazole
SMIL CC1=C(C)C(NC2=C3C=CC=C2)=C3C(OC)=C1OC
Tekniske data
Kemisk navn eller materiale Carbazomycin A
Mængde 50 mg
InChI formel InChI=1S/C16H17NO2/c1-9-10(2)15(18-3)16(19-4)13-11-7-5-6-8-12(11)17-14(9)13/h5-8,17H,1-4H3
Formel vægt 255.1259