missing translation for 'onlineSavingsMsg'
Learn More

Carbasalate calcium, MedChemExpress

Product Code. 16487326
100 mg, 100mg
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MedChemExpress Carbasalate calcium is an anti-inflammatory, antipyretic, and analgesic agent.

Carbasalate calcium is an anti-inflammatory, antipyretic, and analgesic agent.

Specifications

Chemical Name or Material Carbasalate calcium
CAS 5749-67-7
Color White
Purity Grade Notes Research
Molecular Formula C9H7O4.CH4N2O.1/2Ca
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (385.74 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (19.29 mM; Need ultrasonic)
SMILES O=C([O-])C1=CC=CC=C1OC(C)=O.NC(N)=O.[0.5Ca2+]
Molecular Weight (g/mol) 259.24
Formula Weight 259.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Carbasalate calcium, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.