Få mere at vide
Carbaryl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Carbaryl |
| CAS | 63-25-2 |
| Molekylær formel | C12 H11 N O2 |
| Mængde | 25 g |
| Synonym | 1-Naphthalenol, methylcarbamate (9CI), Carbamic acid, methyl-, 1-naphthyl ester (6CI,7CI,8CI), 1-Naphthol N-methylcarbamate, 1-Naphthyl N-methylcarbamate, 1-Naphthyl methylcarbamate, Adios, Agrex S, Arilat, Arilate, Arylam, Atoxan, Bugmaster, Caprolin, Carbaril, Carbaryl, Carbaryl 4L, Carbatox, Carbatox 60, Carbatox 75, Carbavur, Carbomate, Carvin, Carylderm, Clean Crop Sevin 5 Bait, Clinicide, Compound 7744, Denapon, Derbac, Dicarbam, Dicarbam 50, Dyna-carbyl, ENT 23969, Experimental Insecticide 7744, Garden Tech Sevin Concentrate Bug Killer, Garden Tech Sevin Lawn Insect Granules, Garden Tech Sevin Lawn Insecticide, Germain's, Hexavin, Karbatox, Karbatox 75, Karbatox zawiesinowy, Karbosep, Laivin, Laivin 85, Menaphtam, Monsur, Mugan, Murvin, NAC, NAC (insecticide), NMC 50, NSC 27311, OMS 29, Oltitox, Panam, Pomex, Prosevor, Prosevor 85, Ravyon, SA 50 Mole Cricket Bait, Sanol 50, Savite, Seffein, Sevimol, Sevin, Sevin 4, Sevin 4L, Sevin 50WP, Sevin 75, Sevin S 50, Sevin XLR, Sevin XLR plus, Sevin bait, Sevin-4-Oil, Sevnol, Sewin, Slam, Suvamil, Suvamil 50, Suvavil L, Thinsec, UC 7744, Union Carbide 7744, Vetox, Vetox 85, Vioxan, XLR-Plus, α-Naphthalenyl methylcarbamate, α-Naphthyl methylcarbamate |
| InChI formel | InChI=1S/C12H11NO2/c1-13-12(14)15-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3,(H,13,14) |
| SMIL | CNC(=O)Oc1cccc2ccccc12 |
| IUPAC navn | naphthalen-1-yl N-methylcarbamate |
| Molekylvægt (g/mol) | 201.22 |
| Formel vægt | 201.079 |