Få mere at vide
Carbaryl-d7 (naphthyl-d7), CDN
Artikelnummer. 30322055
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322055 0.01 g
30310651 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322055 Leverandør CDN Leverandørnr. CDND54680.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbaryl-d7
CAS 362049-56-7
Molekylær formel C12 2H7 H4 N O2
Mængde 0.01 g
Synonym 1-Naphthalen-2,3,4,5,6,7,8-d7-ol, methylcarbamate (9 CI), 1-Naphthalenyl-2,3,4,5,6,7,8-d7 N-methylcarbamate
InChI formel InChI=1 S/C12H11NO2/c1-13-12(14)15-11-8-4-6-9-5-2-3-7-10(9)11/h2-8 H,1H3,(H,13,14)/i2D,3 D,4 D,5 D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(OC(=O)NC)c([2 H])c([2 H])c([2 H])c2c1[2 H]
IUPAC navn (2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl) N-methylcarbamate
Molekylvægt (g/mol) 208.26
Formel vægt 208.1229 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity