Få mere at vide
(Carbamoylamino)(diphenyl)acetic Acid, Mikromol™
Beskrivelse
(Carbamoylamino)(diphenyl)acetic Acid
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (Carbamoylamino)(diphenyl)acetic Acid |
| CAS | 6802-95-5 |
| Analyt- eller komponentnavne | (Carbamoylamino)(diphenyl)acetic Acid |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H14 N2 O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Phenytoin Imp. E (EP), Phenytoin Sodium Imp. E (EP), Phenytoin USP Related Compound B, Phenytoin USP RC B, (Carbamoylamino)-(diphenyl)acetic Acid, Phenytoin Related Compound B, Phenytoin Impurity E, Phenytoin Sodium Impurity E, Phenytoin Impurity E |
| InChI formel | InChI=1S/C15H14N2O3/c16-14(20)17-15(13(18)19,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H,18,19)(H3,16,17,20) |
| SMIL | O=C(O)C(NC(=O)N)(C=1C=CC=CC1)C=2C=CC=CC2 |
| IUPAC navn | 2,2-diphenyl-2-ureido-acetic acid |
| Vis mere |