Få mere at vide
Carbamazepine-d2 (Major), TRC
Artikelnummer. 30360896
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30360896 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360896 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17584310MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbamazepine-d2
CAS 1189902-21-3
Renhedsgrad noter HPLC
Molekylær formel C15H10D2N2O
Mængde 10 mg
Synonym 5H-Dibenz[b,f]azepine-5-carboxamide-d2, 5-Carbamoyl-5H-dibenz[b,f]azepine-d2, Amizepin-d2, Biston-d2, CBZ-d2, Calepsin-d2, Carbamazepen-d2, Carbamazepin-d2, G 32883-d2, Geigy 32883-d2, Karbamazepin-d2, Karbelex-d2, Karberol-d2, NSC 169864-d2, Tegretol-d2
InChI formel InChI=1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)/i9D,10D
SMIL NC(N(C1=C2C=CC=C1)C3=C(C([2H])=C2[2H])C=CC=C3)=O
IUPAC navn 5,6-dideuteriobenzo[b][1]benzazepine-11-carboxamide
Molekylvægt (g/mol) 238.28
Formel vægt 238.1075
Procent renhed >95