Få mere at vide
Carbamazepine 10,11-Epoxide-d2 (Major), TRC
Artikelnummer. 30360520
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30360520 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360520 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1758531MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbamazepine 10,11-Epoxide-d2
CAS 1185025-41-5
Molekylær formel C15H10D2N2O2
Mængde 1 mg
Synonym 1a,10b-Dihdyro-6H-dibenzo[b,f]oxireno[d]azepine-6-carboxamide-d2, 10,11-Dihydro-10,11-epoxycarbamazepine-d2, 10,11-Epoxycarbamazepine-d2, Carbamazepine-d2 10,11-Oxide, GP 49-023-d2
InChI formel InChI=1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)/i13D,14D
SMIL [2H]C12C([2H])(O2)C3=C(C=CC=C3)N(C(N)=O)C4=C1C=CC=C4
IUPAC navn 2,4-dideuterio-3-oxa-11-azatetracyclo[10.4.0.02,4.05,10]hexadeca-1(16),5,7,9,12,14-hexaene-11-carboxamide
Molekylvægt (g/mol) 254.28
Formel vægt 254.1024