Få mere at vide
Carbadox-d3, TRC
Artikelnummer. 30360690
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30360690 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360690 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17582710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carbadox-d3
CAS 1185240-06-5
Renhedsgrad noter HPLC
Molekylær formel C11H7D3N4O4
Mængde 10 mg
InChI formel InChI=1S/C11H10N4O4/c1-19-11(16)13-12-6-8-7-14(17)9-4-2-3-5-10(9)15(8)18/h2-7H,1H3,(H,13,16)/b12-6-/i1D3
SMIL [2H]C([2H])([2H])OC(=O)N\N=C\c1c[n+]([O-])c2ccccc2[n+]1[O-]
IUPAC navn trideuteriomethyl N-[(Z)-(1,4-dioxidoquinoxaline-1,4-diium-2-yl)methylideneamino]carbamate
Molekylvægt (g/mol) 265.24
Procent renhed >95